Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CTSV | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001106814.4 | XP_001106814.3 | 235 | 141 | ||
XM_028835374.1 | XP_028691207.1 | 123 | 141 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |