Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FABP7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001257714.1 | NP_001244643.1 | 457 | 137 | ||
GenBank mRNA | JU322666.1 | - | 457 | 137 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |