Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GTF2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266870.1 | NP_001253799.1 | 178 | 73 | ||
XM_028827272.1 | XP_028683105.1 | 632 | 73 | |||
GenBank mRNA | JU334178.1 | - | 153 | 73 | ||
JU475311.1 | - | 151 | 73 | |||
JV046860.1 | - | 178 | 73 | |||
JV579683.1 | - | 245 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Macaca mulatta |