Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HAS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015124713.2 | XP_014980199.2 | 2642 | 158 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Macaca mulatta |