Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RUNX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001265962.1 | NP_001252891.1 | 814 | 81 | ||
XM_015132890.2 | XP_014988376.1 | 807 | 81 | |||
XM_028844818.1 | XP_028700651.1 | 1061 | 81 | |||
XM_028844819.1 | XP_028700652.1 | 818 | 81 | |||
GenBank mRNA | JU471230.1 | - | 814 | 81 | ||
JU471231.1 | - | 814 | 81 | |||
JU471232.1 | - | 814 | 81 | |||
JU477609.1 | - | 775 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Macaca mulatta Inventoried |