Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001260929.2 | NP_001247858.1 | 583 | 128 | ||
GenBank mRNA | JU473159.1 | - | 609 | 128 | ||
JV045324.1 | - | 583 | 128 | |||
JV637013.1 | - | 667 | 128 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Macaca mulatta |