Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GZMK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266310.1 | NP_001253239.1 | 668 | 104 | ||
GenBank mRNA | JU475348.1 | - | 668 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Macaca mulatta |