Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ITPKB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015115115.2 | XP_014970601.2 | 3174 | 101 | ||
GenBank mRNA | JU343091.1 | - | 2312 | 101 | ||
JU475499.1 | - | 2958 | 101 | |||
JV505728.1 | - | 1316 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Macaca mulatta |