Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MTAP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001261737.1 | NP_001248666.1 | 853 | 121 | ||
GenBank mRNA | JU334602.1 | - | 849 | 121 | ||
JU475672.1 | - | 836 | 121 | |||
JV047118.1 | - | 853 | 121 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Macaca mulatta |