Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079696.1 | NP_001073164.1 | 1308 | 141 | ||
XM_015132841.2 | XP_014988327.1 | 1485 | 141 | |||
XM_015132842.2 | XP_014988328.1 | 1460 | 141 | |||
XM_015132843.2 | XP_014988329.1 | 1416 | 141 | |||
GenBank mRNA | EF101562.1 | - | 1308 | 141 | ||
JU475682.1 | - | 1357 | 141 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Macaca mulatta |