Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NGFR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001090039.4 | XP_001090039.3 | 936 | 115 | ||
GenBank mRNA | AY738616.1 | - | 817 | 115 | ||
JV652463.1 | - | 462 | 115 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |