Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TNFRSF11B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001096915.3 | XP_001096915.1 | 1004 | 111 | ||
GenBank mRNA | JU327472.1 | - | 580 | 111 | ||
JU487438.1 | - | 126 | 111 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |