Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SERPINB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_028838169.1 | XP_028694002.1 | 417 | 88 | ||
XM_028838170.1 | XP_028694003.1 | 471 | 88 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Macaca mulatta Inventoried |