Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPP1CB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001260652.2 | NP_001247581.1 | 842 | 117 | ||
XM_015111926.2 | XP_014967412.1 | 1233 | 117 | |||
GenBank mRNA | JU335184.1 | - | 670 | 117 | ||
JU335185.1 | - | 670 | 117 | |||
JU335186.1 | - | 670 | 117 | |||
JU335187.1 | - | 670 | 117 | |||
JU471793.1 | - | 668 | 117 | |||
JU471794.1 | - | 724 | 117 | |||
JU471795.1 | - | 724 | 117 | |||
JV043567.1 | - | 838 | 117 | |||
JV047480.1 | - | 842 | 117 | |||
JV047481.1 | - | 842 | 117 | |||
JV529556.1 | - | 793 | 117 | |||
JV654851.1 | - | 681 | 117 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |