Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC106992273 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015128612.2 | XP_014984098.1 | 705 | 101 | ||
XM_015128613.2 | XP_014984099.1 | 706 | 101 | |||
XM_015128614.2 | XP_014984100.1 | 710 | 101 | |||
GenBank mRNA | FJ527018.1 | - | 74 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Recommended Assay Macaca mulatta |