Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSMB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001257425.1 | NP_001244354.1 | 549 | 100 | ||
GenBank mRNA | JU320972.1 | - | 549 | 100 | ||
JU472901.1 | - | 562 | 100 | |||
JV045803.1 | - | 537 | 100 | |||
JV490744.1 | - | 495 | 100 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Macaca mulatta |