Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SFTPC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001042759.1 | NP_001036224.1 | 214 | 71 | ||
GenBank mRNA | U06694.1 | - | 214 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Macaca mulatta |