Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC16A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001257704.1 | NP_001244633.1 | 1481 | 111 | ||
XM_015147143.2 | XP_015002629.2 | 1406 | 111 | |||
XM_015147153.2 | XP_015002639.2 | 2062 | 111 | |||
GenBank mRNA | JU323021.1 | - | 1481 | 111 | ||
JV721619.1 | - | 1489 | 111 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Macaca mulatta |