Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNAPC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001257672.1 | NP_001244601.1 | 152 | 134 | ||
GenBank mRNA | JU336035.1 | - | 152 | 134 | ||
JU476901.1 | - | 174 | 134 | |||
JV048012.1 | - | 152 | 134 | |||
JV659489.1 | - | 197 | 134 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |