Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNCG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266560.1 | NP_001253489.1 | 536 | 116 | ||
XM_028853290.1 | XP_028709123.1 | 618 | 116 | |||
XM_028853291.1 | XP_028709124.1 | 503 | 116 | |||
XM_028853292.1 | XP_028709125.1 | 503 | 116 | |||
XM_028853293.1 | XP_028709126.1 | 503 | 116 | |||
GenBank mRNA | JU323203.1 | - | 536 | 116 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Macaca mulatta |