Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SORD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001260778.1 | NP_001247707.1 | 987 | 123 | ||
GenBank mRNA | JU323287.1 | - | 1060 | 123 | ||
JU476912.1 | - | 987 | 123 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Macaca mulatta |