Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TULP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266799.1 | NP_001253728.1 | 447 | 79 | ||
XM_028828925.1 | XP_028684758.1 | 567 | 79 | |||
GenBank mRNA | JV636353.1 | - | 447 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Macaca mulatta |