Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBE2G2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194170.2 | NP_001181099.1 | 188 | 112 | ||
GenBank mRNA | JU336210.1 | - | 139 | 112 | ||
JV568112.1 | - | 164 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |