Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WNT9B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015119964.2 | XP_014975450.1 | 2762 | 152 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |