Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF84 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015109771.2 | XP_014965257.1 | 753 | 112 | ||
XM_015109772.2 | XP_014965258.1 | 609 | 112 | |||
XM_015109773.2 | XP_014965259.1 | 687 | 112 | |||
XM_028830246.1 | XP_028686079.1 | 768 | 112 | |||
XM_028830247.1 | XP_028686080.1 | 555 | 112 | |||
XM_028830248.1 | XP_028686081.1 | 699 | 112 | |||
GenBank mRNA | JU493010.1 | - | 465 | 112 | ||
JV070639.1 | - | 505 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Macaca mulatta |