Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KMT2D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015151647.2 | XP_015007133.2 | 2092 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |