Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SCARF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015118184.2 | XP_014973670.2 | 1621 | 98 | ||
XM_028835852.1 | XP_028691685.1 | 1621 | 98 | |||
GenBank mRNA | JV664237.1 | - | 332 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Macaca mulatta |