Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HYAL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001260524.1 | NP_001247453.1 | 1042 | 113 | ||
XM_015130783.2 | XP_014986269.1 | 1282 | 113 | |||
XM_015130784.2 | XP_014986270.1 | 1390 | 113 | |||
GenBank mRNA | JU326206.1 | - | 832 | 113 | ||
JV044937.1 | - | 1042 | 113 | |||
JV636573.1 | - | 1147 | 113 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |