Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TNFRSF14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001043357.1 | NP_001036822.1 | 724 | 146 | ||
GenBank mRNA | DQ851094.1 | - | 724 | 146 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Macaca mulatta |