Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBE2M | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194143.2 | NP_001181072.1 | 1058 | 116 | ||
GenBank mRNA | JU472964.1 | - | 437 | 116 | ||
JV632821.1 | - | 440 | 116 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Macaca mulatta |