Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CSRP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001257285.1 | NP_001244214.1 | 327 | 150 | ||
XM_015118823.2 | XP_014974309.2 | 801 | 150 | |||
XM_015118831.2 | XP_014974317.1 | 392 | 150 | |||
XM_028843664.1 | XP_028699497.1 | 452 | 150 | |||
GenBank mRNA | BQ807632.1 | - | 327 | 150 | ||
BQ807767.1 | - | 151 | 150 | |||
JU474399.1 | - | 357 | 150 | |||
JV673835.1 | - | 347 | 150 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |