Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STOM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266229.1 | NP_001253158.1 | 110 | 105 | ||
GenBank mRNA | JU333342.1 | - | 138 | 105 | ||
JU474642.1 | - | 138 | 105 | |||
JV045309.1 | - | 110 | 105 | |||
JV565288.1 | - | 142 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |