Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SERPINB9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266993.1 | NP_001253922.1 | 424 | 107 | ||
XM_015135410.2 | XP_014990896.1 | 426 | 107 | |||
XM_015135411.2 | XP_014990897.1 | 544 | 107 | |||
XM_015135412.2 | XP_014990898.1 | 616 | 107 | |||
GenBank mRNA | JU472759.1 | - | 359 | 107 | ||
JV043268.1 | - | 424 | 107 | |||
JV653736.1 | - | 319 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 Recommended Assay Macaca mulatta Inventoried |