Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NEURL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001113989.4 | XP_001113989.1 | 925 | 118 | ||
XM_015148094.2 | XP_015003580.1 | 688 | 118 | |||
XM_028826812.1 | XP_028682645.1 | 459 | 118 | |||
XM_028826813.1 | XP_028682646.1 | 257 | 118 | |||
GenBank mRNA | JV668151.1 | - | 732 | 118 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |