Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAB3D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194269.2 | NP_001181198.1 | 734 | 124 | ||
GenBank mRNA | JU321236.1 | - | 698 | 124 | ||
JU473643.1 | - | 684 | 124 | |||
JV045771.1 | - | 661 | 124 | |||
JV637107.1 | - | 702 | 124 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Macaca mulatta |