Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NFE2L3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001094650.4 | XP_001094650.2 | 1090 | 124 | ||
GenBank mRNA | JU496783.1 | - | 139 | 124 | ||
JU496784.1 | - | 139 | 124 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |