Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CAMP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033509.2 | NP_001028681.1 | 209 | 138 | ||
GenBank mRNA | AF181954.1 | - | 212 | 138 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |