Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD81 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266249.1 | NP_001253178.1 | 93 | 91 | ||
XM_015113356.2 | XP_014968842.1 | 268 | 91 | |||
XM_015113357.2 | XP_014968843.1 | 268 | 91 | |||
GenBank mRNA | JU337304.1 | - | 136 | 91 | ||
JU470589.1 | - | 93 | 91 | |||
JV600375.1 | - | 281 | 91 | |||
JV637159.1 | - | 1431 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Recommended Assay Macaca mulatta |