Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EFNB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001261375.1 | NP_001248304.1 | 1083 | 63 | ||
GenBank mRNA | JU474587.1 | - | 1178 | 63 | ||
JV043851.1 | - | 1083 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Macaca mulatta |