Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARHGAP35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_028839122.1 | XP_028694955.1 | 4559 | 120 | ||
GenBank mRNA | JU334158.1 | - | 4041 | 120 | ||
JU475285.1 | - | 4048 | 120 | |||
JV046843.1 | - | 4046 | 120 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |