Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCNB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001261149.1 | NP_001248078.1 | 869 | 81 | ||
GenBank mRNA | JU477687.1 | - | 869 | 81 | ||
JU477688.1 | - | 857 | 81 | |||
JV048609.1 | - | 851 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Macaca mulatta |