Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001261124.1 | NP_001248053.1 | 642 | 93 | ||
XM_028827245.1 | XP_028683078.1 | 1475 | 93 | |||
GenBank mRNA | JU472572.1 | - | 642 | 93 | ||
JU472573.1 | - | 642 | 93 | |||
JU472574.1 | - | 642 | 93 | |||
JU472575.1 | - | 642 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Macaca mulatta |