Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: USP38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001260711.1 | NP_001247640.1 | 1748 | 115 | ||
GenBank mRNA | JU336759.1 | - | 2118 | 115 | ||
JU477523.1 | - | 1748 | 115 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |