Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPINK7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015141206.2 | XP_014996692.1 | 146 | 138 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Macaca mulatta |