Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLEKHA8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001199076.2 | NP_001186005.2 | 783 | 142 | ||
GenBank mRNA | JU473730.1 | - | 458 | 142 | ||
JV704143.1 | - | 466 | 142 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |