Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AOPEP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015118087.2 | XP_014973573.2 | 2602 | 123 | ||
XM_015118088.2 | XP_014973574.2 | 2602 | 123 | |||
XM_015118094.2 | XP_014973580.2 | 2305 | 123 | |||
XM_028835410.1 | XP_028691243.1 | 2806 | 123 | |||
XM_028835411.1 | XP_028691244.1 | 2806 | 123 | |||
GenBank mRNA | JV073828.1 | - | 331 | 123 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Macaca mulatta |