Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PGAP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001265734.1 | NP_001252663.1 | 516 | 122 | ||
GenBank mRNA | JV636459.1 | - | 516 | 122 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |