Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PGAP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001265734.1 | NP_001252663.1 | 915 | 71 | ||
XM_015119517.2 | XP_014975003.1 | 718 | 71 | |||
GenBank mRNA | JV636459.1 | - | 915 | 71 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Macaca mulatta |