Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SELENOI | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001135563.3 | NP_001129035.1 | 464 | 61 | ||
GenBank mRNA | JU477593.1 | - | 415 | 61 | ||
JV596838.1 | - | 100 | 61 | |||
JV725791.1 | - | 443 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Macaca mulatta |