Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLIC6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001087698.4 | XP_001087698.2 | 1752 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Macaca mulatta |